Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP11A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299558PCP
Description
ARHGAP11A Polyclonal antibody specifically detects ARHGAP11A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ARHGAP11A | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
GAP (1-12), KIAA0013, MGC70740, Rho GTPase activating protein 11A, rho GTPase-activating protein 11A, rho-type GTPase-activating protein 11A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ARHGAP11A. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
9824 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction