Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546085 Supplier Invitrogen™ Supplier No. PA546085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MLGIDGLCAT PSLEGFEEGE YETPGEYKRK RRQSVGDFVS GALNKFKPNR Sequence homology: Cow: 93%; Dog: 77%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 86%; Rabbit: 92%; Rat: 93%.

ARHGAP11A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP11A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARHGAP11A
Gene Accession No. Q6P4F7
Gene Alias 6530401L14Rik; Arhgap11a; GAP (1-12); Kiaa0013; mKIAA0013; RGD1309107; Rho GTPase activating protein 11A; rho GTPase-activating protein 11A; Rho-type GTPase-activating protein 11A
Gene Symbols ARHGAP11A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ARHGAP11A (aa 252-301).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9824
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.