Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP11A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580348
Description
This antibody has specificity for Human ARHGAP11A.
ARHGAP11A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include signal transduction.
Specifications
ARHGAP11A | |
Polyclonal | |
Unconjugated | |
ARHGAP11A | |
6530401L14Rik; Arhgap11a; GAP (1-12); Kiaa0013; mKIAA0013; RGD1309107; Rho GTPase activating protein 11A; rho GTPase-activating protein 11A; Rho-type GTPase-activating protein 11A | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
9824 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q6P4F7 | |
ARHGAP11A | |
A synthetic peptide corresponding to the center region of the Human ARHGAP11A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction