Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP11B Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306142PE
Description
ARHGAP11B Polyclonal antibody specifically detects ARHGAP11B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ARHGAP11B | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
B'-T, FAM7B1, Family With Sequence Similarity 7, Member B1, GAP (1-8), Rho GTPase Activating Protein 11B, Rho GTPase-Activating Protein 11B, Rho-Type GTPase-Activating Protein 11B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ARHGAP11B. | |
0.1 mL | |
GPCR, Signal Transduction | |
89839 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction