Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP11B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106949 Supplier Invitrogen™ Supplier No. PA5106949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 85%; Dog: 77%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Rabbit: 83%; Rat: 85%.

ARHGAP11B is a Hominin-specific protein that promotes development and evolutionary expansion of the brain neocortex. Able to promote amplification of basal progenitors in the subventricular zone, producing more neurons during fetal corticogenesis. Does not possess GTPase activator activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP11B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene ARHGAP11B
Gene Accession No. Q3KRB8
Gene Alias ARHGAP11B; B'-T; FAM7B1; family with sequence similarity 7, member B1; GAP (1-8); Protein FAM7B1; Rho GTPase activating protein 11B; rho GTPase-activating protein 11B; Rho-type GTPase-activating protein 11B
Gene Symbols ARHGAP11B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human ARHGAP11B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.