Learn More
ARHGAP12 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304740AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.Specifications
ARHGAP12 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8C0D4, Q8IWW6 | |
Arhgap12 | |
Between 100 and 150 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ARHGAP12 | |
FLJ10971, FLJ20737, FLJ21785, rho-type GTPase-activating protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
75415, 94134 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.