Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531530 Supplier Invitrogen™ Supplier No. PA531530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat. Predicted reactivity: Mouse (87%), Rat (82%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RHO GTPases regulate diverse biologic processes, and their activity is regulated by RHO GTPase-activating proteins (GAPs), such as ARHGAP15 (Seoh et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ARHGAP15
Gene Accession No. Q53QZ3, Q5ZMM3
Gene Alias 5830480G12Rik; ArhGAP15; BM-024; BM-030; BM046; BM-046; hypothetical protein; polymorphic marker clone XE09; RCJMB04_1j23; Rho GTPase activating protein 15; rho GTPase-activating protein 15; Rho-type GTPase-activating protein 15
Gene Symbols ARHGAP15
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARHGAP15. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 424303, 55843
Target Species Chicken, Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.