Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982303
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2303 Supplier Bioss Supplier No. BS12519R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RHO GTPases regulate diverse biologic processes, and their activity is regulated by RHO GTPase-activating proteins (GAPs), such as ARHGAP15 (Seoh et al., 2003 ).

Specifications

Antigen ARHGAP15
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARHGAP15
Gene Accession No. Q53QZ3
Gene Alias 5830480G12Rik; ARHGAP15; BM-024; BM-030; BM046; BM-046; BOS_1779; polymorphic marker clone XE09; RCJMB04_1j23; Rho GTPase activating protein 15; Rho GTPase-activating protein 15; Rho-type GTPase-activating protein 15
Gene Symbols ARHGAP15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARHGAP15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55843
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.