Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145117 Supplier Invitrogen™ Supplier No. PA5145117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPKPRNRPSV PPPPQPPGVH SAGDSSLTNT APTASKIVTD SNSRVSEPHR SIFPEMHSDS ASKDVPGRIL L Antigen sequence identity: Mouse 75%; Rat 75%.

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA (MIM 165390), RAC1 (MIM 602048), and CDC42 (MIM 116952).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP17
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Arhgap17
Gene Accession No. Q68EM7
Gene Alias 5730403H17Rik; Arhgap17; MST066; MST110; MSTP038; MSTP066; MSTP110; Nadrin; Nadrin2; neuron-associated developmentally regulated protein; neuron-associated developmentally-regulated protein; neuron-specific GTPase activating protein; neuron-specific GTPase-activating protein; PP367; PP4534; Rho GTPase activating protein 17; rho GTPase-activating protein 17; RhoGAP interacting with CIP4 homologs 1; RhoGAP interacting with CIP4 homologs protein 1; Rho-type GTPase-activating protein 17; Rich1; RICH-1; RICH1B; WBP15; WW domain-binding protein 15
Gene Symbols Arhgap17
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 800-870. Recombinant protein control fragment (Product #RP-110214).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.