Learn More
ARHGAP17 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304689A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA (MIM 165390), RAC1 (MIM 602048), and CDC42 (MIM 116952).Specifications
ARHGAP17 | |
Polyclonal | |
Unconjugated | |
ARHGAP17 | |
MST066, MST110, MSTP038, MSTP066, MSTP110, NADRIN, PP367, PP4534, RICH1, RICH1B, WBP15, RICH-1, RhoGAP interacting with CIP4 homologs 1, neuron-associated developmentally regulated protein, rho GTPase-activating protein 17, rho-type GTPase-activating protein 17, rhoGAP interacting with CIP4 homologs protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55114 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q68EM7 | |
ARHGAP17 | |
Between 757 and 625 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.