Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP18 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281704F
This item is not returnable.
View return policy
Description
ARHGAP18 Polyclonal antibody specifically detects ARHGAP18 in Human, Mouse, Rat samples. It is validated for ELISA, ImmunofluorescenceSpecifications
ARHGAP18 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunofluorescence | |
FITC | |
bA307O14.2, MacGAP, MGC126757, Rho GTPase activating protein 18, rho-type GTPase-activating protein 18 | |
ARHGAP18 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ARHGAP18. The immunogen is located within the last 50 amino acids of ARHGAP18. Amino Acid Squence: DTYMKDLYQLNPNAE | |
0.1 mL | |
Signal Transduction | |
93663 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction