Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP18 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281704H
This item is not returnable.
View return policy
Description
ARHGAP18 Polyclonal antibody specifically detects ARHGAP18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
ARHGAP18 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
HRP | |
bA307O14.2, MacGAP, MGC126757, Rho GTPase activating protein 18, rho-type GTPase-activating protein 18 | |
ARHGAP18 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ARHGAP18. The immunogen is located within the last 50 amino acids of ARHGAP18. Amino Acid Squence: DTYMKDLYQLNPNAE | |
0.1 mL | |
Signal Transduction | |
93663 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction