Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545144 Supplier Invitrogen™ Supplier No. PA545144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PTPESVAIGE LKGTSKENRN LLFSGSPAVT MTPTRLKWSE GKKEGKKGFL Sequence homology: Cow: 91%; Dog: 100%; Human: 100%; Mouse: 77%; Rabbit: 100%; Rat: 83%.

Members of the ARHGAP family, such as ARHGAP19, encode negative regulators of Rho GTPases (see RHOA; MIM 165390), which are involved in cell migration, proliferation, and differentiation, actin remodeling, and G1 cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARHGAP19
Gene Accession No. Q14CB8
Gene Alias 4933411B03Rik; AI120128; ARHGAP19; AW546831; mFLJ00368; putative RhoGAP protein; Rho GTPase activating protein 19; rho GTPase-activating protein 19; Rho-type GTPase-activating protein 19
Gene Symbols ARHGAP19
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ARHGAP19 (aa 445-494).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84986
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.