Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560285 Supplier Invitrogen™ Supplier No. PA560285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLTHKHFNAH LKIADLMQFD DKGNKTNIPD KDRQIEALQL LFLILPPPNR NLLKLLLDLL YQTAKKQDKN KMSAYNLALM F Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Members of the ARHGAP family, such as ARHGAP19, encode negative regulators of Rho GTPases (see RHOA; MIM 165390), which are involved in cell migration, proliferation, and differentiation, actin remodeling, and G1 cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP19
Gene Accession No. Q14CB8
Gene Alias 4933411B03Rik; AI120128; ARHGAP19; AW546831; mFLJ00368; putative RhoGAP protein; Rho GTPase activating protein 19; rho GTPase-activating protein 19; Rho-type GTPase-activating protein 19
Gene Symbols ARHGAP19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGAP19. Recombinant protein control fragment (Product #RP-97231).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.