Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP19 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310823100UL
Description
ARHGAP19 Polyclonal specifically detects ARHGAP19 in Human samples. It is validated for Western Blot.Specifications
ARHGAP19 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
DKFZp313K217, MGC138804, MGC138805, MGC14258, putative RhoGAP protein, Rho GTPase activating protein 19, rho GTPase-activating protein 19, rho-type GTPase-activating protein 19 | |
The immunogen is a synthetic peptide directed towards the middle region of human ARHGAP19 (NP_001191229.1). Peptide sequence HFNAHLKIADLMQFDDKGNKTNIPDKDRQIEALQLLFLILPPPNRNLLKL | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
84986 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction