Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP181411
Description
ARHGAP20 Polyclonal specifically detects ARHGAP20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGAP20 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9P2F6 | |
ARHGAP20 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KGPRTHRRCSEPNIEDQNRKLTYLRGIYSKKQHKTSCEAGLLHGEEDYLKRHKSLQMEGQKLINQSLVMGIEVGKSSATNQNTE | |
0.1 mL | |
Signal Transduction | |
57569 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
KIAA1391Rho-type GTPase-activating protein 20, RA and RhoGAP domain containing protein, RARHOGAP, Rho GTPase activating protein 20, rho GTPase-activating protein 20 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction