Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558398 Supplier Invitrogen™ Supplier No. PA558398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGPRTHRRCS EPNIEDQNRK LTYLRGIYSK KQHKTSCEAG LLHGEEDYLK RHKSLQMEGQ KLINQSLVMG IEVGKSSATN QNTE Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 86%.

ARHGAP20 is a potent GTPase-activating protein (GAP) and convert Rho-type GTPases into an inactive GDP-bound state. It contains a pleckstrin homology (PH) domain, a Ras-associating domain, a Rho-GAP domain and two Annexin-like (ANXL) repeats. ARHGAP20 is a direct downstream target of Rap1 in the neurite outgrowth while it's over expression leads to inactivation of Rho for promoting the neurite outgrowth in a Rap1-dependent manner. ARHGAP20 is a tumor suppressor gene and is inactivated by deletion in breast cancer and also by chromosomal translocation in B-cell chronic lymphocytic leukemia. Reports suggest that it may take part in rearrangements of the cytoskeleton and cell signaling events that occur during spermatogenesis. It is expressed predominantly in human brain, liver, ovary and spinal cord while low expression is found in lymph nodes and fetal liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP20
Gene Accession No. Q9P2F6
Gene Alias 6530403F17Rik; A530023E23Rik; Arhgap20; KIAA1391; mKIAA1391; RA and RhoGAP domain containing protein; RA and RhoGAP domain-containing protein; RARhoGAP; Rho GTPase activating protein 20; rho GTPase activating protein 20 variant 2; rho GTPase-activating protein 20; RhoGTPase activating protein; Rho-type GTPase-activating protein 20
Gene Symbols ARHGAP20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGAP20. Recombinant protein control fragment (Product #RP-97366).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57569
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.