Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100388 Supplier Invitrogen™ Supplier No. PA5100388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARHGAP10.

Functions as a GTPase-activating protein (GAP) for RHOA and CDC42. Downstream partner of ARF1 which may control Golgi apparatus structure and function. Also required for CTNNA1 recruitment to adherens junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP21
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP21
Gene Accession No. Q5T5U3, Q6DFV3
Gene Alias 5530401C11Rik; AA416458; ARHGAP10; ARHGAP21; Kiaa1424; Rho GTPase activating protein 21; rho GTPase-activating protein 10; Rho GTPase-activating protein 21; Rho-GTPase activating protein 10; rho-type GTPase-activating protein 21
Gene Symbols ARHGAP21
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP21(Accession Q5T5U3), corresponding to amino acid residues A416-V466.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57584, 71435
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.