Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557742 Supplier Invitrogen™ Supplier No. PA557742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPPCLTTSAP LIRRQLSHDH ESVGPPSLDA QPNSKTERSK SYDEGLDDYR EDAKLSFKHV SSLKGIKIAD SQKSSEDSGS RKDSSSEVFS DAAKEG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.

Functions as a GTPase-activating protein (GAP) for RHOA and CDC42. Downstream partner of ARF1 which may control Golgi apparatus structure and function. Also required for CTNNA1 recruitment to adherens junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP21
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP21
Gene Accession No. Q5T5U3
Gene Alias 5530401C11Rik; AA416458; ARHGAP10; ARHGAP21; Kiaa1424; Rho GTPase activating protein 21; rho GTPase-activating protein 10; Rho GTPase-activating protein 21; Rho-GTPase activating protein 10; rho-type GTPase-activating protein 21
Gene Symbols ARHGAP21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGAP21. Recombinant protein control fragment (Product #RP-97044).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57584
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.