Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595800 Supplier Invitrogen™ Supplier No. PA595800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVPAPGLVPG IPSVASMAWS GASSSESSVG GSLSSCTACR ASDSSARSSL HTDWALEPSP LPSSSEDPKS LDLDHSMDEA GAGASNSEPS EPDSPTREHA RRSEALQGLV TELRAELCRQ RTEYERSVKR IEEGSADLRK R; Positive Samples: Mouse kidney, Mouse lung, Rat kidney, Rat lung; Cellular Location: Cytoplasm, Nucleus.

GTPase-activating proteins (GAPs) accelerate the intrinsic rate of GTP hydrolysis of Ras-related proteins, resulting in down regulation of their active form. ARHGAP22 (Rho GTPase activating protein 22), also known as RHOGAP2, is a 698 amino acid protein that localizes to the cytoplasm, and contains one Rho-GAP domain and one PH domain. ARHGAP22 functions as a GTPase-activating protein for Rac 1 converting it to an inactive GDP-bound state and, through its interaction with ZNF161 (VEZF1), is thought to be useful in transcription regulation. Multiple isoforms of ARHGAP22 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP22
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Arhgap22
Gene Accession No. Q8BL80
Gene Alias Arhgap22; B230341L19Rik; p68RacGAP; RHG22; Rho GTPase activating protein 22; rho GTPase-activating protein 22; RHOGAP2; RhoGap22; Rho-type GTPase-activating protein 22
Gene Symbols Arhgap22
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 510-650 of human ARHGAP22 (NP_0012429531).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239027, 306279
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.