Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | ARHGAP24 |
---|---|
Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ARHGAP24 Polyclonal specifically detects ARHGAP24 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGAP24 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp564B1162, Filamin-A-associated RhoGAP, FilGAP, FILGAPRCGAP72, FLJ33877, p73, p73RhoGAP, RAC1- and CDC42-specific GTPase-activating protein of 72 kDa, RC-GAP72, Rho GTPase activating protein 24, rho GTPase-activating protein 24, RhoGAP of 73 kDa, Rho-type GTPase-activating protein 24, Sarcoma antigen NY-SAR-88 | |
ARHGAP24 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q8N264 | |
83478 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ILNSDTLGNPTNVRNMSWLPNGYVTLRDNKQKEQAGELGQHNRLSTYDNVHQQFSMMNLDDKQSIDSATWSTSSCEISLPENSNSCRSSTTTCPEQDFFGGNFEDPVLDGPPQDDLSHPRDYESKSD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title