Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104104 Supplier Invitrogen™ Supplier No. PA5104104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHG24.

Rho GTPase-activating protein involved in cell polarity, cell morphology and cytoskeletal organization. Acts as a GTPase activator for the Rac-type GTPase by converting it to an inactive GDP-bound state. Controls actin remodeling by inactivating Rac downstream of Rho leading to suppress leading edge protrusion and promotes cell retraction to achieve cellular polarity. Able to suppress RAC1 and CDC42 activity in vitro. Overexpression induces cell rounding with partial or complete disruption of actin stress fibers and formation of membrane ruffles, lamellipodia, and filopodia. Isoform 2 is a vascular cell-specific GAP involved in modulation of angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP24
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP24
Gene Accession No. Q5U2Z7, Q8C4V1, Q8N264
Gene Alias 0610025G21Rik; Arhgap24; Down-regulated in nephrectomized rat kidney #2; DR-NR#2; Filamin-A-associated RhoGAP; FILGAP; p73; p73RhoGAP; RAC1- and CDC42-specific GTPase-activating protein of 72 kDa; RCGAP72; RC-GAP72; Rho GTPase activating protein 24; rho GTPase-activating protein 24; rhoGAP of 73 kDa; Rho-type GTPase-activating protein 24; Sarcoma antigen NY-SAR-88; si:ch211-65i12.2
Gene Symbols ARHGAP24
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP24(Accession Q8N264), corresponding to amino acid residues S573-A623.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231532, 305156, 83478
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.