Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986801
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6801 Supplier Bioss Supplier No. BS5785R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rho GTPase-activating protein involved in cell polarity, cell morphology and cytoskeletal organization. Acts as a GTPase activator for the Rac-type GTPase by converting it to an inactive GDP-bound state. Controls actin remodeling by inactivating Rac downstream of Rho leading to suppress leading edge protrusion and promotes cell retraction to achieve cellular polarity. Able to suppress RAC1 and CDC42 activity in vitro. Overexpression induces cell rounding with partial or complete disruption of actin stress fibers and formation of membrane ruffles, lamellipodia, and filopodia. Isoform 2 is a vascular cell-specific GAP involved in modulation of angiogenesis.

Specifications

Antigen ARHGAP24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARHGAP24
Gene Accession No. Q8N264
Gene Alias 0610025G21Rik; ARHGAP24; Down-regulated in nephrectomized rat kidney #2; DR-NR#2; Filamin-A-associated RhoGAP; FILGAP; p73; p73RhoGAP; RAC1- and CDC42-specific GTPase-activating protein of 72 kDa; RCGAP72; RC-GAP72; Rho GTPase activating protein 24; Rho GTPase-activating protein 24; RhoGAP of 73 kDa; Rho-type GTPase-activating protein 24; Sarcoma antigen NY-SAR-88
Gene Symbols ARHGAP24
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARHGAP24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83478
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.