Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588434 Supplier Invitrogen™ Supplier No. PA588434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLGQSACLF LSIARSRSVM TGEQMAAFHP SSTPNPLERP IKMGWLKKQR SIVKNWQQRY FVLRAQQLYY YKDEEDTKPQ GCMYLPGCTI KEIATNPEEA GKFVFEIIPA SWDQNRMGQD SYVLMASSQA EMEEWVKFLR RVAGTPCGAV FGQRLDETVA YEQKFGPHLV PILVEKCAEF ILEHGRNEEG IFRLPGQDNL VKQLRDAFDA GERPSFDRDT DVHTVASLLK LYLRDLPEPV VPWSQYEGFL LCGQLTNADE AKAQQELMKQ LSILPRDNYS LLSYICRFLH EIQLNCAVNK MSVDNLATVI GVNLIRSKVE DPAVIMRGTP QIQRVMTMMI RDHEVLFPKS KDIPLSPPAQ KNDPKKAPVA; Positive Samples: COS-7, OVCAR3.

ARHGAP25 encode negative regulators of Rho GTPases, which are implicated in actin remodeling, cell polarity, and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP25
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARHGAP25
Gene Accession No. P42331, Q8BYW1
Gene Alias A130039I20Rik; ARHGAP25; epididymis secretory protein Li 308; HEL-S-308; KAIA0053; KIAA0053; RGD1562105; Rho GTPase activating protein 25; rho GTPase-activating protein 25; Rho-type GTPase-activating protein 25
Gene Symbols ARHGAP25
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232201, 9938
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.