Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556464 Supplier Invitrogen™ Supplier No. PA556464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQSIRDVRDI FGVSESPPRD TCGNHTNQLD GTKEERELPR VIKTSGSMPD DASLNSTTLS DASQDKEGSF AVPRSDSVAI LETIP Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 67%.

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP28
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP28
Gene Accession No. Q9P2N2
Gene Alias ARHGAP28; AU044757; AW550892; E130310N06; Kiaa1314; RGD1559882; Rho GTPase activating protein 28; rho GTPase-activating protein 28; Rho-type GTPase-activating protein 28
Gene Symbols ARHGAP28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGAP28. Recombinant protein control fragment (Product #RP-95582).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.