Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556465
Description
Immunogen sequence: KSIPRCRRIN RMLSNESLHP PAFSRSNSEA SVDSASMEDF WREIESIKDS SMGGQEEPPP AEVTPVD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
ARHGAP28 | |
Polyclonal | |
Unconjugated | |
ARHGAP28 | |
ARHGAP28; AU044757; AW550892; E130310N06; Kiaa1314; RGD1559882; Rho GTPase activating protein 28; rho GTPase-activating protein 28; Rho-type GTPase-activating protein 28 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2N2 | |
ARHGAP28 | |
Recombinant protein corresponding to Human ARHGAP28. Recombinant protein control fragment (Product #RP-95583). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction