Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115842 Supplier Invitrogen™ Supplier No. PA5115842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHG28.

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP28
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ARHGAP28
Gene Accession No. Q8BN58, Q9P2N2
Gene Alias ARHGAP28; AU044757; AW550892; E130310N06; Kiaa1314; RGD1559882; Rho GTPase activating protein 28; rho GTPase-activating protein 28; Rho-type GTPase-activating protein 28
Gene Symbols ARHGAP28
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP28(Accession Q9P2N2), corresponding to amino acid residues N278-G328.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268970, 301709, 79822
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.