Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116779
Description
Positive Samples: Mouse spleen Immunogen sequence: PLADSGPDDL APALEDSLSQ EVQDSFSFLE DSSSSEPEWV GAEDGEVAQA EAAGAAFSPG EDDPGMGYLE ELLGVGPQVE EFSVEPPLDD LSLDEAQFVL A.
ARHGAP30 is GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
ARHGAP30 | |
Polyclonal | |
Unconjugated | |
ARHGAP30 | |
6030405P05Rik; ARHGAP30; Gm102; mFLJ00267; RGD1561419; Rho GTPase activating protein 30; rho GTPase-activating protein 30; Rho-type GTPase-activating protein 30 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
226652, 257106 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.73 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q640N3, Q7Z6I6 | |
ARHGAP30 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 485-585 of human ARHGAP30 (NP_8590712). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction