Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP30 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31555620UL
Description
ARHGAP30 Polyclonal antibody specifically detects ARHGAP30 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ARHGAP30 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rho GTPase activating protein 30 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 485-585 of human ARHGAP30 (NP_859071.2). | |
20 μg | |
GPCR, Signal Transduction | |
257106 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction