Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144353
Description
ARHGAP32 Polyclonal Antibody for ICC/IF, ELISA
GTPase-activating protein (GAP) promoting GTP hydrolysis on RHOA, CDC42 and RAC1 small GTPases. May be involved in the differentiation of neuronal cells during the formation of neurite extensions. Involved in NMDA receptor activity-dependent actin reorganization in dendritic spines. May mediate cross-talks between Ras- and Rho regulated signaling pathways in cell growth regulation. Isoform 2 has higher GAP activity.
Specifications
ARHGAP32 | |
Polyclonal | |
Unconjugated | |
ARHGAP32 | |
3426406O18Rik; 6430596G11Rik; AI841135; ARHGAP32; Arhgap35; brain-specific Rho GTP-ase-activating protein; brain-specific Rho GTPase-activating protein; BRF2; butyrate response factor 2; EGF-response factor 2; ERF2; ERF-2; GAB-associated CDC42; GAB-associated Cdc42/Rac GTPase-activating protein; GAP-associated protein p190; Gc-gap; glucocorticoid receptor DNA binding factor 1; glucocorticoid receptor DNA-binding factor 1; Glucocorticoid receptor repression factor 1; GRF1; GRF-1; Grit; Grlf1; GTPase regulator interacting with TrkA; GTPase-activating protein for Cdc42 and Rac1; KIAA0712; KIAA1722; mKIAA0712; mKIAA1722; mRNA decay activator protein ZFP36L2; P190 RhoGAP; p190A; p190-A; p190ARHOGAP; p190RhoGAP; p200RhoGAP; p250Gap; PX-RICS; rac GTPase activating protein; rho GAP p190A; Rho GTPase activating protein 32; Rho GTPase activating protein 33 pseudogene; Rho GTPase activating protein 35; rho GTPase-activating protein 32; Rho GTPase-activating protein 35; Rho/Cdc42/Rac GTPase-activating protein RICS; rhoGAP involved in the beta-catenin-N-cadherin and NMDA receptor signaling; RhoGAP involved in the -catenin-N-cadherin and NMDA receptor signaling; rho-type GTPase-activating protein 32; RICS; RNF162C; TIS11D; TPA-induced sequence 11d; ZFP36 ring finger protein like 2; ZFP36 ring finger protein-like 2; Zfp36l2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2 | |
Rabbit | |
Protein G | |
RUO | |
9743 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
2.565 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
A7KAX9 | |
ARHGAP32 | |
Recombinant Human Rho GTPase-activating protein 32 protein (130-205AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction