Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107063
Description
Sequence homology: Cow: 93%; Dog: 93%; Goat: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Pig: 86%; Sheep: 93%.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
| ARHGAP36 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP36 | |
| ARHGAP36; Rho GTPase activating protein 36; rho GTPase-activating protein 36 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 158763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q6ZRI8 | |
| ARHGAP36 | |
| Synthetic peptide directed towards the middle region of human RP13-102H20.1 (aa 468-518). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction