Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP42 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558989
Description
Immunogen sequence: DPSIPLPQPQ SRSGSRRTRA ICLSTGSRKP RGRYTPCLAE PDSDSYSSSP DSTPMGSIES LSSHSSEQNS TTKSASCQ Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
This gene encodes a Rho GTPase-activating protein (RhoGAP), and member of the GRAF or BAR-PH family of proteins. Expression of this gene is enriched in vascular smooth muscle cells and the encoded protein inhibits RhoA activity to regulate vascular tone and control blood pressure. A mutation in the first intron of this gene modulates its expression and is associated with reduced blood pressure in human patients with borderline hypertension.
Specifications
ARHGAP42 | |
Polyclonal | |
Unconjugated | |
ARHGAP42 | |
9030420J04Rik; Arhgap42; BE136619; Graf3; RGD1564081; Rho GTPase activating protein 42; Rho GTPase-activating protein 10-like; Rho GTPase-activating protein 42; rho-type GTPase-activating protein 42; TMEM133; VRho GTPase activating protein 42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
143872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NI28 | |
ARHGAP42 | |
Recombinant protein corresponding to Human ARHGAP42. Recombinant protein control fragment (Product #RP-97422). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction