Learn More
ARHGAP5 Mouse anti-Human, Clone: OTI9H2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809437

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Rho GTPase activating protein 5 negatively regulates RHO GTPases, a family which may mediate cytoskeleton changes by stimulating the hydrolysis of bound GTP. Two transcript variants encoding different isoforms have been found for this gene. GTPase-activating protein for Rho family members. May play a role in the reduction of the p21rasGTPase-activating potential of RASA1/p120GAP.Specifications
ARHGAP5 | |
Monoclonal | |
Unconjugated | |
ARHGAP5 | |
GFI2, RhoGAP5, p190-B, p190BRhoGAP | |
Mouse | |
Affinity Chromatography | |
RUO | |
394 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9H2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13017 | |
ARHGAP5 | |
Human recombit protein fragment corresponding to amino acids 948-1261 of human ARHGAP5 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.