Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP5 Recombinant Rabbit Monoclonal Antibody (3L9I3)
GREENER_CHOICE

Catalog No. PIMA538043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538043 Supplier Invitrogen™ Supplier No. MA538043
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, MCF7, C2C12, Mouse brain, Rat brain Immunogen sequence: PSKYKYKSKT LFSKAKSYYR RTHSDASDDE AFTTSKTKRK GRHRGSEEDP LLSPVETWKG GIDNPAITSD QELDDKKMKK KTHKVKEDKK QKKKTKNFNP PTRRNWESNY FGMPLQDLVT AEKPIPLFVE KCVEFIEDTG LCTEGLYRVS G.

Rho GTPase activating protein 5 negatively regulates RHO GTPases, a family which may mediate cytoskeleton changes by stimulating the hydrolysis of bound GTP. Two transcript variants encoding different isoforms have been found for this gene. GTPase-activating protein for Rho family members. May play a role in the reduction of the p21rasGTPase-activating potential of RASA1/p120GAP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP5
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 3L9I3
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARHGAP5
Gene Accession No. P97393, Q13017
Gene Alias ARHGAP5; GFI2; growth factor independent 2; LRRGT00098; p100 RasGAP-associated p105 protein; p105 RhoGAP; p190B; p190-B; p190BRhoGAP; Rho GTPase activating protein 5; rho GTPase-activating protein 5; Rho GTPase-activating protein 5-like protein; RHOGAP5; rho-type GTPase-activating protein 5
Gene Symbols ARHGAP5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1150-1300 of human RhoGAP (Q13017).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11855, 299012, 394
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.