Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ARHGAP8 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ARHGAP8 Polyclonal specifically detects ARHGAP8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ARHGAP8 | |
Polyclonal | |
Rabbit | |
Human | |
23779 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AYKEFDRKDGDLTMWPRLVS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
BCH Domain-Containing Cdc42GAP-Like Protein, BNIP-2 And Cdc42GAP Homology Domain-Containing, Proline-Rich And BPGAP1, Cdc42GAP-Like Protein Subtype-1, PP610, Rho GTPase Activating Protein 8, Rho GTPase-Activating Protein 8, Rho-Type GTPase-Activating Protein 8 | |
ARHGAP8 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title