Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGAP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570690
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 93%.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
ARHGAP8 | |
Polyclonal | |
Unconjugated | |
ARHGAP8 | |
3110043J09Rik; ARHGAP8; BCH domain-containing Cdc42GAP-like protein; BNIP-2 and Cdc42GAP homology domain-containing, proline-rich and Cdc42GAP-like protein subtype-1; BPGAP1; PP610; Rho GTPase activating protein 8; rho GTPase-activating protein 8; Rho-type GTPase-activating protein 8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23779 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P85298 | |
ARHGAP8 | |
synthetic peptide directed towards the middle region of human ARHGAP8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction