Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567065 Supplier Invitrogen™ Supplier No. PA567065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYKEFDRKDGD LTMWPRLVS Highest antigen sequence identity to the following orthologs - mouse 40%, rat 45%.

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP8
Gene Accession No. P85298
Gene Alias 3110043J09Rik; ARHGAP8; BCH domain-containing Cdc42GAP-like protein; BNIP-2 and Cdc42GAP homology domain-containing, proline-rich and Cdc42GAP-like protein subtype-1; BPGAP1; PP610; Rho GTPase activating protein 8; rho GTPase-activating protein 8; Rho-type GTPase-activating protein 8
Gene Symbols ARHGAP8
Host Species Rabbit
Immunogen Recombinant Human ARHGAP8. Recombinant protein control fragment (Product #RP-105483).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23779
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.