Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP9 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ARHGAP9 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0388
![]() ![]() |
Proteintech Group Inc
156651AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the Rho-GAP family of GTPase activating proteins. The protein has substantial GAP activity towards several Rho-family GTPases in vitro, converting them to an inactive GDP-bound state. It is implicated in regulating adhesion of hematopoietic cells to the extracellular matrix. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
ARHGAP9 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9BRR9 | |
64333 | |
ARHGAP9 Fusion Protein Ag8104 | |
Primary | |
-20°C | |
ARHGAP9 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
10C, ARHGAP9, RGL1 | |
ARHGAP9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title