Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGDIA Monoclonal Antibody (OTI1F2), TrueMAB™, OriGene
SDP

Catalog No. 502542545
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2545 30 μL
50-167-2233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2545 Supplier OriGene Supplier No. TA501089S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARHGDIA Monoclonal antibody specifically detects ARHGDIA in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen ARHGDIA
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DAPK2
Gene Accession No. Q9UIK4
Gene Alias 5330430M07Rik; Arhgdia; arhgdia {ECO:0000312; Bles01; C87222; epididymis secretory sperm binding protein Li 47e; Gdi1; Gdi-1; GDIA1; GDP-dissociation inhibitor, aplysia RAS-related 1; HEL-S-47e; MGC117248; NPHS8; QtrA-11619; RGD:1359547}; Rho GDI 1; rho GDI 1 {ECO:0000250; Rho GDI alpha; Rho GDP dissociation inhibitor (GDI) alpha; Rho GDP dissociation inhibitor alpha; rho GDP-dissociation inhibitor 1; rho GDP-dissociation inhibitor 1 {ECO:0000250; RhoDGI; RHOGDI; Rho-GDI; Rho-GDI alpha; rho-GDI alpha {ECO:0000250; RhoGDI-1; UniProtKB:P19803}; unnamed protein product
Host Species Mouse
Immunogen Full length human recombinant protein of human DAPK2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23604
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.