Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146831UV
Description
ARHGEF1 Polyclonal antibody specifically detects ARHGEF1 in Human samples. It is validated for Western BlotSpecifications
ARHGEF1 | |
Polyclonal | |
Western Blot | |
115 kDa guanine nucleotide exchange factor, 115-kD protein, GEF1, LBCL2LSC, Lsc homolog, p115RhoGEF, P115-RHOGEF, Rho guanine nucleotide exchange factor (GEF) 1, rho guanine nucleotide exchange factor 1, Sub1.5 | |
The immunogen for this product maps to a region between residue 775 and 825 of human 115 kDa Rho guanine nucleotide exchange factor using the numbering given in entry NP_004697.2 (GeneID 9138). | |
0.1 mL | |
Cell Cycle and Replication | |
9138 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction