Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGEF10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101845
Description
Antibody detects endogenous levels of total ARHGEF10.
This gene encodes a Rho guanine nucleotide exchange factor (GEF). Rho GEFs regulate the activity of small Rho GTPases by stimulating the exchange of guanine diphosphate (GDP) for guanine triphosphate (GTP) and may play a role in neural morphogenesis. Mutations in this gene are associated with slowed nerve conduction velocity (SNCV). Alternative splicing results in multiple transcript variants.
Specifications
ARHGEF10 | |
Polyclonal | |
Unconjugated | |
ARHGEF10 | |
6430549H08Rik; Arhgef10; GEF10; Kiaa0294; mKIAA0294; RGD1565043; Rho guanine nucleotide exchange factor (GEF) 10; Rho guanine nucleotide exchange factor 10; SNCV | |
Rabbit | |
Affinity chromatography | |
RUO | |
9639 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15013 | |
ARHGEF10 | |
A synthesized peptide derived from human ARHGEF10(Accession O15013), corresponding to amino acid residues L338-S388. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction