Learn More
ARHGEF10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303308AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ARHGEF10 (rho guanine nucleotide exchange factor 10), also known as GEF10 or KIAA0294, is a 1,369 amino acid protein that contains one DBL-homology domain and is thought to play a role in myelination of peripheral nerves, specifically during development. ARHGEF10 is expressed as four alternatively spliced isoforms that are present at low levels in ovary, lung, testis and kidney, with considerably higher expression in the central and peripheral nervous systems. Defects in the gene encoding ARHGEF10 are the cause of slowed nerve conduction velocity (SNCV), an autosomal domit disorders that is characterized by a reduction in nerve velocities resulting from lack of proper myelination.Specifications
ARHGEF10 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O15013 | |
ARHGEF10 | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ARHGEF10 | |
Rho guanine nucleotide exchange factor 10, ARHGEF10, KIAA0294 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9639 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.