Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539417
Description
ARHGEF10 (rho guanine nucleotide exchange factor 10), also known as GEF10 or KIAA0294, is a 1,369 amino acid protein that contains one DBL-homology domain and is thought to play a role in myelination of peripheral nerves, specifically during development. ARHGEF10 is expressed as four alternatively spliced isoforms that are present at low levels in ovary, lung, testis and kidney, with considerably higher expression in the central and peripheral nervous systems. Defects in the gene encoding ARHGEF10 are the cause of slowed nerve conduction velocity (SNCV), an autosomal domit disorders that is characterized by a reduction in nerve velocities resulting from lack of proper myelination.
Specifications
ARHGEF10 | |
Polyclonal | |
Unconjugated | |
ARHGEF10 | |
Rho guanine nucleotide exchange factor 10, ARHGEF10, KIAA0294 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9639 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15013 | |
ARHGEF10 | |
A synthetic peptide derived from the internal region of human ARHGEF10 | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction