Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $402.00
Specifications
Antigen | ARHGEF11 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.1-0.5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224517SS
![]() |
Novus Biologicals
NBP224517SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224517
![]() |
Novus Biologicals
NBP224517 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
ARHGEF11 Polyclonal specifically detects ARHGEF11 in Human, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGEF11 | |
Western Blot 0.1-0.5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Rabbit | |
Human, Rat, Bovine, Primate | |
O15085 | |
9826 | |
A portion of amino acids 1-50 of human ARHGEF11 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
DKFZp667F1223, glutamate transporter EAAT4-associated protein 48, GTRAP48, KIAA0380rho guanine nucleotide exchange factor 11, PDZ-RhoGEF, Rho guanine exchange factor (GEF) 11, Rho guanine nucleotide exchange factor (GEF) 11, RhoA-specific guanine nucleotide exchange factor, RhoGEF glutamate transport modulator | |
ARHGEF11 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), rat, mouse, dog and chimpanzee. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title