Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP183314
Description
ARHGEF11 Polyclonal specifically detects ARHGEF11 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGEF11 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
DKFZp667F1223, glutamate transporter EAAT4-associated protein 48, GTRAP48, KIAA0380rho guanine nucleotide exchange factor 11, PDZ-RhoGEF, Rho guanine exchange factor (GEF) 11, Rho guanine nucleotide exchange factor (GEF) 11, RhoA-specific guanine nucleotide exchange factor, RhoGEF glutamate transport modulator | |
Rabbit | |
Affinity Purified | |
RUO | |
9826 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ARHGEF11 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LQAEIDSRLRNSEDARGVLCEAQEAAMPEIQEQIHDYRTKRTLGLGSLYGENDLLDLDGDPLRERQVAEKQLAALGDILSKYEEDRSAPMDFALNTYMSHAGIRLREARPSNTAEKAQSAPDKDKWLPFFPKTKKSSNSKKEQDA | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction