Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552897 Supplier Invitrogen™ Supplier No. PA552897
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGHTMETQAA QEPEDDLTPT PSVISVTSHP WDPGSPGQAP PGGEGDNTQL AGLEGERPEQ EDMGLCSLEH LPPRTRNSGI WESPELDRNL AEDASSTEAA GGYKVVRKAE VAGSKVVPAL P Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 62%.

ARHGEF11 is a 1527 amino acid protein with tandem dbl homology and pleckstrin homology domains, PDZ domain, two proline rich sequences and a regulatory G protein-signaling (RGS) sequence. It acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase, but not for RAC1 or CDC42 and acts as GTPase-activating protein (GAP) for GNA12 and GNA13. It forms a complex with G proteins and stimulates neurite through retraction Rho-dependent signals. ARHGEF11 increases glutamate transport 4-fold. It interacts with EAAT4 and modulates its uptake activity and perisynaptic distribution at glutamatergic synapses. It is known to induce the reorganization of the actin cytoskeleton and its overexpression induces the formation of membrane ruffling and filopodia. It is ubiquitously expressed, in brain, lung, liver, kidney and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF11
Gene Accession No. O15085
Gene Alias Arhgef11; B930073M02; E130307F09; glutamate transporter EAAT4 associated protein 48; glutamate transporter EAAT4-associated protein 48; Gtrap48; KIAA0380; mKIAA0380; PDZ-RhoGEF; Prg; Rho guanine exchange factor (GEF) 11; Rho guanine nucleotide exchange factor (GEF) 11; rho guanine nucleotide exchange factor 11; RhoA-specific guanine nucleotide exchange factor; RhoGEF; RhoGEF glutamate transport modulator; RhoGEF glutamate transport modulator GTRAP48; RP11-356J7.2
Gene Symbols ARHGEF11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGEF11. Recombinant protein control fragment (Product #RP-89159).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.