Learn More
Invitrogen™ ARHGEF11 Polyclonal Antibody

Description
Immunogen sequence: FIICTSKLGP PQIYELVALT SSDKNTWMEL LEEAVRNATR HPGAAPMPVH PPPPGPREPA QQGPTPSRVE LDDSDVFHGE PEPEELPGGT GSQQRVQGKH QVLLEDPEQE GSAEEEELGV LPCPSTSLDG ENRGIR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 68%.
Specifications
Specifications
| Antigen | ARHGEF11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ARHGEF11 |
| Gene Accession No. | O15085 |
| Gene Alias | Arhgef11; B930073M02; E130307F09; glutamate transporter EAAT4 associated protein 48; glutamate transporter EAAT4-associated protein 48; Gtrap48; KIAA0380; mKIAA0380; PDZ-RhoGEF; Prg; Rho guanine exchange factor (GEF) 11; Rho guanine nucleotide exchange factor (GEF) 11; rho guanine nucleotide exchange factor 11; RhoA-specific guanine nucleotide exchange factor; RhoGEF; RhoGEF glutamate transport modulator; RhoGEF glutamate transport modulator GTRAP48; RP11-356J7.2 |
| Gene Symbols | ARHGEF11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.