Learn More
Invitrogen™ ARHGEF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143957
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEL whole cell, human K562 whole cell, rat brain tissue. Flow: HL-60 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli working through G protein-coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein has been observed to form a myeloid/lymphoid fusion partner in acute myeloid leukemia. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| ARHGEF12 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF12 | |
| 2310014B11Rik; Arhgef12; AU019857; KIAA0382; LARG; leukemia-associated Rho guanine nucleotide exchange factor; leukemia-associated Rho guanine-nucleotide exchange factor; leukemia-associated RhoGEF; mKIAA0382; PRO2792; Rho guanine nucleotide exchange factor (GEF) 12; Rho guanine nucleotide exchange factor 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23365, 367072 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q9NZN5 | |
| ARHGEF12 | |
| E.coli-derived human LARG/ARHGEF12 recombinant protein (Position: T4-S254). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.