Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101847 Supplier Invitrogen™ Supplier No. PA5101847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARHGEF12.

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli working through G protein-coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein has been observed to form a myeloid/lymphoid fusion partner in acute myeloid leukemia. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGEF12
Gene Accession No. Q8R4H2, Q9NZN5
Gene Alias 2310014B11Rik; Arhgef12; AU019857; KIAA0382; LARG; leukemia-associated Rho guanine nucleotide exchange factor; leukemia-associated Rho guanine-nucleotide exchange factor; leukemia-associated RhoGEF; mKIAA0382; PRO2792; Rho guanine nucleotide exchange factor (GEF) 12; Rho guanine nucleotide exchange factor 12
Gene Symbols ARHGEF12
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGEF12(Accession Q9NZN5), corresponding to amino acid residues K583-P633.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23365, 367072, 69632
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.