Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106723 Supplier Invitrogen™ Supplier No. PA5106723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARHGEF16.

Although the specific function of this protein is not known yet, it is thought to be involved in protein-protein and protein-lipid interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF16
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Arhgef16
Gene Accession No. Q5VV41
Gene Alias Arhgef16; EPHEXIN4; Ephexin-4; GEF16; NBR; Neuroblastoma; Rho guanine exchange factor (GEF) 16; Rho guanine nucleotide exchange factor (GEF) 16; Rho guanine nucleotide exchange factor 16
Gene Symbols Arhgef16
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGEF16(Accession Q5VV41), corresponding to amino acid residues G166-Y216.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27237
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.